missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenylcarbazide, ACS, Thermo Scientific Chemicals

387.00 SEK - 2645.00 SEK

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.282
MDL Number MFCD00003013
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11367458
View Documents
Thermo Scientific Alfa Aesar
031390.06
5 g
387.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11377458
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031390.14
25 g
950.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
11387458
View Documents
Thermo Scientific Alfa Aesar
031390.22
100 g
2645.00 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1,5-Diphenylcarbazide is used in the colorimetric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

140-22-7
242.282
KSPIHGBHKVISFI-UHFFFAOYSA-N
8789
1,3-dianilinourea
C13H14N4O
MFCD00003013
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
CHEBI:4641
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Specifications

140-22-7
White to Cream
MFCD00003013
Air and light sensitive
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
KSPIHGBHKVISFI-UHFFFAOYSA-N
1,3-dianilinourea
8789
242.28
Crystalline
170°C to 176°C
C13H14N4O
752039
14,3321
Soluble in glacial acetic acid,alcohol and acetone; Slightly soluble in water.
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
242.282
CHEBI:4641
ACS
1,5-Diphenylcarbazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 205-403-7

missing translation for 'rtecsNumber' : FF2750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • 1,5-Diphenylcarbazide

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.