missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 60°C to 65°C |
Color | White to Yellow |
Boiling Point | 127°C to 128°C (2.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 25 g |
Beilstein | 03,786E |
Show More |
Safety and Handling

Product Identifier
- 1-Benzyl-4-hydroxypiperidine
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction