missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Boc-3-azetidinemethanol, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
500 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an intermediate in organic syntheses
Solubility
Soluble in Dimethyl sulfoxide (DMSO).
Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
It is an intermediate in organic syntheses
Solubility
Soluble in Dimethyl sulfoxide (DMSO).
Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
Specifications
Specifications
Boiling Point | 268°C to 273°C |
Synonym | tert-butyl 3-hydroxymethyl azetidine-1-carboxylate, 1-boc-azetidine-3-yl-methanol, 1-boc-azetidine-3-yl methanol, 1-boc-azetidine-3-ylmethanol, 1-boc-3-hydroxymethyl azetidine, 1-boc-3-azetidinemethanol, 1-tert-butoxycarbonyl-3-azetidinemethanol, 1-azetidinecarboxylic acid, 3-hydroxymethyl-, 1,1-dimethylethyl ester, tert-butyl3-hydroxymethyl azetidine-1-carboxylate, 3-hydroxymethyl-azetidine-1-carboxylic acid tert-butyl ester |
Solubility Information | Soluble in Dimethyl sulfoxide (DMSO). |
Formula Weight | 187.24 |
Percent Purity | 95% |
Density | 1.115 |
Quantity | 500 mg |
Flash Point | 117°C (243°F) |
Chemical Name or Material | 1-Boc-3-azetidinemethanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'