Learn More
1-Boc-6-cyanoindole-2-boronic acid, 96%, Thermo Scientific Chemicals
1245.00 SEK - 3835.00 SEK
Chemical Identifiers
CAS | 913835-67-3 |
---|---|
Molecular Formula | C14H15BN2O4 |
Molecular Weight (g/mol) | 286.09 |
MDL Number | MFCD08436056 |
InChI Key | CZEBTZMIQZVUSE-UHFFFAOYSA-N |
Synonym | 1-boc-6-cyanoindole-2-boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indole-2-boronic acid, 6-cyano-1h-indole-2-boronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyanoindol-2-ylboronic acid, acmc-209r9t, 6-cyano-1h-indol-2-ylboronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronicacid, 1-tert-butoxycarbonyl-2-dihydroxyboryl-1h-indole-6-carbonitrile |
PubChem CID | 44119347 |
IUPAC Name | [6-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid |
SMILES | CC(C)(C)OC(=O)N1C(=CC2=CC=C(C=C12)C#N)B(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489257
|
Thermo Scientific Alfa Aesar
H52520.MD |
250 mg |
1245.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15479257
|
Thermo Scientific Alfa Aesar
H52520.03 |
1 g |
3835.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
913835-67-3 | |
286.09 | |
CZEBTZMIQZVUSE-UHFFFAOYSA-N | |
44119347 | |
CC(C)(C)OC(=O)N1C(=CC2=CC=C(C=C12)C#N)B(O)O |
C14H15BN2O4 | |
MFCD08436056 | |
1-boc-6-cyanoindole-2-boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indole-2-boronic acid, 6-cyano-1h-indole-2-boronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyanoindol-2-ylboronic acid, acmc-209r9t, 6-cyano-1h-indol-2-ylboronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronicacid, 1-tert-butoxycarbonyl-2-dihydroxyboryl-1h-indole-6-carbonitrile | |
[6-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid |
Specifications
913835-67-3 | |
C14H15BN2O4 | |
1-boc-6-cyanoindole-2-boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-6-cyano-1h-indole-2-boronic acid, 6-cyano-1h-indole-2-boronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyanoindol-2-ylboronic acid, acmc-209r9t, 6-cyano-1h-indol-2-ylboronic acid, n-boc protected, 1-tert-butoxycarbonyl-6-cyano-1h-indol-2-yl boronicacid, 1-tert-butoxycarbonyl-2-dihydroxyboryl-1h-indole-6-carbonitrile | |
CC(C)(C)OC(=O)N1C(=CC2=CC=C(C=C12)C#N)B(O)O | |
286.09 | |
286.09 | |
1-Boc-6-cyanoindole-2-boronic acid |
219°C to 221°C | |
MFCD08436056 | |
CZEBTZMIQZVUSE-UHFFFAOYSA-N | |
[6-cyano-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid | |
44119347 | |
96% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.