Learn More
1-Bromo-2,3,5,6-tetrafluorobenzene, 99%, Thermo Scientific Chemicals
Description
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Specifications
Density | 1.883 |
Boiling Point | 142°C to 144°C |
Flash Point | 53°C (129°F) |
Odor | Aromatic |
Refractive Index | 1.469 |
Quantity | 5 g |
UN Number | UN1993 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 228.98 |
Percent Purity | 99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10240216 | Certificate of Analysis | 21/04/2022 | B24265.06,B24265.14 |
10226292 | Certificate of Analysis | 23/11/2021 | B24265.06,B24265.14 |
10229420 | Certificate of Analysis | 23/11/2021 | B24265.06,B24265.14 |
10155858 | Certificate of Analysis | 23/11/2021 | B24265.06,B24265.14 |
10201726 | Certificate of Analysis | 23/11/2021 | B24265.06,B24265.14 |