missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,5-bis(trifluoromethyl)benzene, 97%, Thermo Scientific™

Product Code. 11487166
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7617-93-8
Molecular Formula C8H3BrF6
Molecular Weight (g/mol) 293.006
MDL Number MFCD03094336
InChI Key GFQNSGHVOFVTLC-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl bromobenzene, 2-bromo-1,4-bis trifluoromethyl benzene, 1-bromo-2,5-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-bromobenzene, benzene, 2-bromo-1,4-bis trifluoromethyl, pubchem4264, acmc-1bisb, ksc495k7j, 2,5-bis-trifluoromethylbromobenzene
PubChem CID 2734763
IUPAC Name 2-bromo-1,4-bis(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)C(F)(F)F

Specifications

Density 1.691
Boiling Point 146°C to 147°C
Flash Point >110°C (230°F)
Refractive Index 1.434
Quantity 1 g
Formula Weight 293.01
Percent Purity 97%
Chemical Name or Material 1-Bromo-2,5-bis(trifluoromethyl)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromo-2,5-bis(trifluoromethyl)benzene, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.