missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-ethylbenzene, 98%, Thermo Scientific Chemicals

Product Code. 11485427
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1973-22-4
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000077
InChI Key HVRUGFJYCAFAAN-UHFFFAOYSA-N
PubChem CID 16089
IUPAC Name 1-bromo-2-ethylbenzene
SMILES CCC1=CC=CC=C1Br

Specifications

Melting Point -68°C
Density 1.338
Boiling Point 199°C to 200°C
Flash Point 71°C (159°F)
Refractive Index 1.5485
Quantity 25 g
Beilstein 1929735
Synonym o-bromoethylbenzene, benzene, 1-bromo-2-ethyl, 2-ethylbromobenzene, 1-ethyl-2-bromobenzene, 2-bromo-1-ethylbenzene, 1-bromo-2-ethyl-benzene, benzene,1-bromo-2-ethyl, o-ethylbromobenzene, 2-bromophenyl ethane, bromo-2-ethylbenzene
Formula Weight 185.06
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-ethylbenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'