Learn More
1-Bromo-4-fluoro-2-iodobenzene, 98+%, Thermo Scientific Chemicals
Product Code. 11470868
Click to view available options
Quantity:
1 g
5 g
25 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 202865-72-3
Molecular Formula C6H3BrFI
Molecular Weight (g/mol) 300.897
MDL Number MFCD00042578
InChI Key QDCBSSUCEUMLGC-UHFFFAOYSA-N
Synonym 2-bromo-5-fluoroiodobenzene, 4-fluoro-2-iodobromobenzene, 2-iodo-4-fluorobromobenzene, 1-bromo-4-fluoro-2-iodo-benzene, benzene, 1-bromo-4-fluoro-2-iodo, 2-bromo-5-fluoro-1-iodobenzene, pubchem3233, acmc-1cljm, ksc493k3j, 1-bromo-2-iodo-4-fluorobenzene
PubChem CID 2773370
IUPAC Name 1-bromo-4-fluoro-2-iodobenzene
SMILES C1=CC(=C(C=C1F)I)Br
Specifications
Density 2.3
Boiling Point 235°C
Flash Point >110°C (230°F)
Refractive Index 1.63
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 300.9
Percent Purity ≥98%
Chemical Name or Material 1-Bromo-4-fluoro-2-iodobenzene