Learn More
(1-Bromoethyl)benzene, 97%, Thermo Scientific Chemicals
673.34 SEK - 1905.24 SEK
Chemical Identifiers
CAS | 585-71-7 |
---|---|
Molecular Formula | C8H9Br |
Molecular Weight (g/mol) | 185.06 |
MDL Number | MFCD00000139 |
InChI Key | CRRUGYDDEMGVDY-UHFFFAOYSA-N |
Synonym | 1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene |
PubChem CID | 11454 |
IUPAC Name | 1-bromoethylbenzene |
SMILES | CC(C1=CC=CC=C1)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10625872
|
Thermo Scientific Acros
218390250 |
25 g | Glass bottle |
673.34 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10753534
|
Thermo Scientific Acros
218391000 |
100 g | Glass bottle |
1905.24 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
585-71-7 | |
185.06 | |
CRRUGYDDEMGVDY-UHFFFAOYSA-N | |
11454 | |
CC(C1=CC=CC=C1)Br |
C8H9Br | |
MFCD00000139 | |
1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene | |
1-bromoethylbenzene |
Specifications
585-71-7 | |
100.0 | |
1.3500g/mL | |
81°C | |
96% min. (GC) | |
C6H5CH(CH3)Br | |
MFCD00000139 | |
1.35 | |
CRRUGYDDEMGVDY-UHFFFAOYSA-N | |
1-bromoethylbenzene | |
11454 | |
97% | |
(1-Bromoethyl)benzene |
96.0 | |
Brown to Yellow | |
94.0°C (16.0 mmHg) | |
Authentic | |
C8H9Br | |
1.5593 to 1.5613 | |
05, 355 | |
1-bromoethyl benzene, 1-phenylethyl bromide, benzene, 1-bromoethyl, 1-bromo-1-phenylethane, 1-phenethyl bromide, 1-phenyl-1-bromoethane, alpha-phenylethyl bromide, alpha-methylbenzyl bromide, alpha-phenethyl bromide, alpha-bromoethyl benzene | |
CC(C1=CC=CC=C1)Br | |
185.06 | |
185.06 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 209-560-2
RUO – Research Use Only