Learn More
1-Chloro-3,5-difluorobenzene, 97%, Thermo Scientific™
Product Code. 11362986
Click to view available options
Quantity:
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1435-43-4
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00041518
InChI Key RFKBODCWHNDUTJ-UHFFFAOYSA-N
Synonym 3,5-difluorochlorobenzene, benzene, 1-chloro-3,5-difluoro, benzene,1-chloro-3,5-difluoro, 1-chloro-3,5-difluoro-benzene, 3,5-difluorochlorobenzene, 5-difluorochlorobenzene, pubchem4318, 3,5-difluorchlorbenzol, acmc-209cqr, intermediates-zcf02092, 1-chloro-3,5difluorobenzene
PubChem CID 137000
IUPAC Name 1-chloro-3,5-difluorobenzene
SMILES C1=C(C=C(C=C1F)Cl)F
Specifications
Density 1.329
Boiling Point 117°C to 119°C
Flash Point 29°C (84°F)
Odor Aromatic
Refractive Index 1.468
Quantity 1 g
UN Number UN1993
Beilstein 2205670
Formula Weight 148.54
Percent Purity 97%
Chemical Name or Material 1-Chloro-3,5-difluorobenzene
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.