missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-N,N,2-trimethylpropenylamine, 98.5+%, Thermo Scientific Chemicals

1573.93 SEK - 6044.80 SEK

Chemical Identifiers

CAS 26189-59-3
Molecular Formula C6H12ClN
Molecular Weight (g/mol) 133.62
InChI Key GQIRIWDEZSKOCN-UHFFFAOYSA-N
Synonym 1-chloro-n,n,2-trimethylpropenylamine, 1-chloro-n,n,2-trimethyl-1-propenylamine, 1-chloro-n,n,2-trimethyl-1-propen-1-amine, ghosez inverted exclamation markas reagent, 1-propen-1-amine, 1-chloro-n,n,2-trimethyl, 1-chloro-2-methylprop-1-en-1-yl dimethylamine, acmc-1ceik, 1-chloro-1-dimethylaminoisobutene, 1-chloro-2,n,n-trimethylpropenylamine, 1-chloro-n,n-2-trimethylpropenylamine
PubChem CID 640792
IUPAC Name 1-chloro-N,N,2-trimethylprop-1-en-1-amine
SMILES CC(=C(N(C)C)Cl)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10217750
SDS View Documents
Thermo Scientific Acros
300270050
5 mL Glass bottle
1573.93 SEK
5mL
Please to purchase this item. Need a web account? Register with us today!
11935181
View Documents
Thermo Scientific Acros
300270250
25 mL Glass bottle
6044.80 SEK
25mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chloro-N,N,2-trimethylpropenylamine, 98.5+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.