Learn More
1-Cyclohexene-1-carbonitrile, 98%, Thermo Scientific Chemicals
724.00 SEK - 2335.00 SEK
Chemical Identifiers
CAS | 1855-63-6 |
---|---|
Molecular Formula | C7H9N |
Molecular Weight (g/mol) | 107.156 |
MDL Number | MFCD00013772 |
InChI Key | GTMWGXABXQTZRJ-UHFFFAOYSA-N |
Synonym | 1-cyanocyclohexene, cyanocyclohexene, 1-cyclohexene-1-carbonitrile, cyclohexenecarbonitrile, 1-cyclohexenecarbonitrile, cyclohex-1-enecarbonitrile, cyclohex-1-ene-1-carbonitrile, 1-cyclohexenyl cyanide, 1-cyano-1-cyclohexene, acmc-1cqlm |
PubChem CID | 74619 |
IUPAC Name | cyclohexene-1-carbonitrile |
SMILES | C1CCC(=CC1)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11308345
|
Thermo Scientific Alfa Aesar
L10284.04 |
2 g |
724.00 SEK
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318345
|
Thermo Scientific Alfa Aesar
L10284.09 |
10 g |
2335.00 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
1855-63-6 | |
107.156 | |
GTMWGXABXQTZRJ-UHFFFAOYSA-N | |
74619 | |
C1CCC(=CC1)C#N |
C7H9N | |
MFCD00013772 | |
1-cyanocyclohexene, cyanocyclohexene, 1-cyclohexene-1-carbonitrile, cyclohexenecarbonitrile, 1-cyclohexenecarbonitrile, cyclohex-1-enecarbonitrile, cyclohex-1-ene-1-carbonitrile, 1-cyclohexenyl cyanide, 1-cyano-1-cyclohexene, acmc-1cqlm | |
cyclohexene-1-carbonitrile |
Specifications
1855-63-6 | |
85°C to 88°C (17 mmHg) | |
C7H9N | |
MFCD00013772 | |
385837 | |
Not miscible in water. | |
C1CCC(=CC1)C#N | |
107.156 | |
107.16 | |
1-Cyclohexene-1-carbonitrile |
0.947 | |
83°C (181°F) | |
1.482 | |
UN3276 | |
1-cyanocyclohexene, cyanocyclohexene, 1-cyclohexene-1-carbonitrile, cyclohexenecarbonitrile, 1-cyclohexenecarbonitrile, cyclohex-1-enecarbonitrile, cyclohex-1-ene-1-carbonitrile, 1-cyclohexenyl cyanide, 1-cyano-1-cyclohexene, acmc-1cqlm | |
GTMWGXABXQTZRJ-UHFFFAOYSA-N | |
cyclohexene-1-carbonitrile | |
74619 | |
98% |
Safety and Handling
GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
H227-H302+H312+H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
missing translation for 'einecsNumber' : 217-454-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 1-Cyclohexene-1-carbonitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Cyclohexenecarbonitrile
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.