missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-1-butanol, 98%, Thermo Scientific™

Product Code. 11450591
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4352-42-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00019360
InChI Key MTUCYAOJXPTLHZ-UHFFFAOYNA-N
Synonym 1-cyclohexyl-1-butanol, cyclohexanemethanol, .alpha.-propyl, alpha-propylcyclohexanemethanol, zyklohexyl-butanol, cyclohexanemethanol, alpha-propyl, acmc-1ctfc, 1-hydroxybutyl cyclohexane, n-propyl cyclohexyl carbinol, mtucyaojxptlhz-uhfffaoysa
PubChem CID 107269
IUPAC Name 1-cyclohexylbutan-1-ol
SMILES CCCC(O)C1CCCCC1

Specifications

Density 0.902
Boiling Point 103°C to 104°C (4 mmHg)
Flash Point 108°C (228°F)
Refractive Index 1.465
Quantity 5 g
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material 1-Cyclohexyl-1-butanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Cyclohexyl-1-butanol, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.