missing translation for 'onlineSavingsMsg'
Learn More
1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals Product Code.: 11411518

1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11411518
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11411518

Brand: Thermo Scientific Alfa Aesar A13852.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.

Solubility
Immiscible in water.

Notes
Avoid excessive heat and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1493-27-2
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD00007048
InChI Key PWKNBLFSJAVFAB-UHFFFAOYSA-N
Synonym 2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitroShow MoreShow Less
PubChem CID 73895
IUPAC Name 1-fluoro-2-nitrobenzene
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])F

Specifications

Melting Point -8°C
Density 1.34
Boiling Point 215°C
Flash Point 94°C (201°F)
Assay Percent Range 99%
Refractive Index 1.5317
Quantity 250 g
UN Number UN2810
Beilstein 607262
Solubility Information Immiscible in water.
Formula Weight 141.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-2-nitrobenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10207568 Certificate TypeCertificate of Analysis Date29/12/2023 Catalog NumberA13852.30,A13852.0B,A13852.18
Lot Number10218981 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13852.30,A13852.0B,A13852.18
Lot Number10227369 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13852.30,A13852.0B,A13852.18
Lot Number10226584 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13852.30,A13852.0B,A13852.18
Lot Number10207568 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13852.30,A13852.0B,A13852.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.