missing translation for 'onlineSavingsMsg'
Learn More
1-Methoxy-2-methyl-2-propanol, 98+%, Thermo Scientific Chemicals Product Code.: 11339196

1-Methoxy-2-methyl-2-propanol, 98+%, Thermo Scientific Chemicals

Product Code. 11339196
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11339196

Brand: Thermo Scientific Alfa Aesar L20351.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3587-64-2
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD03701584
InChI Key MXUXZWFVAPTPAG-UHFFFAOYSA-N
Synonym 1-methoxy-2-methyl-2-propanol, 2-propanol, 1-methoxy-2-methyl, acmc-20aott, 2-methoxymethyl-2-propanol, 1,1-dimethyl-2-methoxyethanol, 2-propanol,1-methoxy-2-methyl, 1-methoxy-2-methyl-2-propanol #Show MoreShow Less
PubChem CID 77137
IUPAC Name 1-methoxy-2-methylpropan-2-ol
SMILES COCC(C)(C)O

Specifications

Density 0.892
Boiling Point 115°C to 116°C
Flash Point 27°C (80°F)
Refractive Index 1.4044
Quantity 25 g
UN Number UN1987
Formula Weight 104.15
Percent Purity ≥98%
Chemical Name or Material 1-Methoxy-2-methyl-2-propanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10178793 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL20351.14,L20351.22
Lot Number10231875 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL20351.14,L20351.22
Lot Number10139928 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL20351.14,L20351.22
Lot Number10178863 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL20351.14,L20351.22
Lot Number10228808 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL20351.14,L20351.22
compliance-icons compliance-icons
Product Identifier
  • 1-Methoxy-2-methyl-2-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Methoxy-2-methyl-2-propanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methoxy-2-methyl-2-propanol, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.