missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-imidazole-4-carbonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber glass bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Synonym | 1-methyl-1h-imidazole-4-carbonitrile, 4-cyano-1-methyl-1h-imidazole, 1h-imidazole-4-carbonitrile, 1-methyl, 4-cyano-1-methylimidazole, 1-methyl-4-cyano-1h-imidazole, 1-methyl-4-imidazolecarbonitrile, 1h-imidazole-4-carbonitrile,1-methyl |
Formula Weight | 107.12 |
Percent Purity | 97% |
Chemical Name or Material | 1-Methyl-1H-imidazole-4-carbonitrile |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-Methyl-1H-imidazole-4-carbonitrile, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction