missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester is used as a pharmaceutical intermediate, agrochemical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester is used as a pharmaceutical intermediate, agrochemical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Specifications
Specifications
Melting Point | 58°C to 61°C |
Quantity | 5 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 208.07 |
Percent Purity | 98% |
Chemical Name or Material | 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 98%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction