Learn More
Tocris Bioscience™ 1-Methyl-D-tryptophan
Indoleamine 2,3-dioxygenase (IDO) inhibitor
Brand:  Tocris Bioscience™ 5698/50
Encompass
Product Code. 15787056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
1-Methyl-D-tryptophan is an indoleamine 2,3-dioxygenase (IDO) inhibitor (IC50 = 7 μM); disrupts tryptophan catabolism. Enhances the antitumor and antiviral immunoresponses of CD8+ T-cells in vitro. Reduces tumor volume in mice with xenografts overexpressing IDO.
Specifications
Specifications
110117-83-4
1MT