Learn More
1-Naphthyl phosphate disodium salt hydrate, 99%, Thermo Scientific Chemicals
1-Naphthyl phosphate disodium salt hydrate, 99%, C10H9Na2O5P, CAS Number-207569-06-0
749.00 SEK - 8980.00 SEK
Chemical Identifiers
CAS | 207569-06-0 |
---|---|
Molecular Formula | C10H7Na2O4P |
Molecular Weight (g/mol) | 268.12 |
MDL Number | MFCD00041007 |
InChI Key | QYURIFWAOPAPAJ-UHFFFAOYSA-L |
Synonym | 1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt |
PubChem CID | 24212014 |
IUPAC Name | disodium;naphthalen-1-yl phosphate;hydrate |
SMILES | [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429917
|
Thermo Scientific Alfa Aesar
B22866.03 |
1 g |
749.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439917
|
Thermo Scientific Alfa Aesar
B22866.06 |
5 g |
2690.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449917
|
Thermo Scientific Alfa Aesar
B22866.14 |
25 g |
8980.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Chemical Identifiers
207569-06-0 | |
268.12 | |
QYURIFWAOPAPAJ-UHFFFAOYSA-L | |
24212014 | |
[Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 |
C10H7Na2O4P | |
MFCD00041007 | |
1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt | |
disodium;naphthalen-1-yl phosphate;hydrate |
Specifications
207569-06-0 | |
C10H7Na2O4P | |
4895811 | |
Soluble in water. | |
[Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 | |
268.12 | |
268.12 (Anhydrous) | |
1-Naphthyl phosphate disodium salt hydrate |
>300°C | |
MFCD00041007 | |
1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt | |
QYURIFWAOPAPAJ-UHFFFAOYSA-L | |
disodium;naphthalen-1-yl phosphate;hydrate | |
24212014 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 218-564-3
missing translation for 'tsca' : No
Recommended Storage : Keep cold
- 1-Naphthyl phosphate disodium salt hydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 1-Naphthylphosphatedisodium salt hydrate