missing translation for 'onlineSavingsMsg'
Learn More

1-(pyrid-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride, 97%, Thermo Scientific™

1554.75 SEK - 4947.97 SEK

Chemical Identifiers

CAS 946409-40-1
Molecular Formula C12H18Cl2N2O2
Molecular Weight (g/mol) 293.188
MDL Number MFCD06801260
InChI Key FLINLLCJYJQIIY-UHFFFAOYSA-N
Synonym 1-pyridin-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride, 1-pyrid-3-ylmethyl piperidine-4-carboxylic acid dihydrochloride, 1-pyridin-3-yl methyl piperidine-4-carboxylic acid dihydrochloride, 1-pyridin-3-yl methyl piperidine-4-carboxylic acid-hydrogen chloride 1/2
PubChem CID 42935493
IUPAC Name 1-(pyridin-3-ylmethyl)piperidine-4-carboxylic acid;dihydrochloride
SMILES C1CN(CCC1C(=O)O)CC2=CN=CC=C2.Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10575944
SDS View Documents
Thermo Scientific Maybridge
CC69851DA
1 g Amber Glass Bottle
1554.75 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10123044
SDS View Documents
Thermo Scientific Maybridge
CC69851DE
5 g Amber Glass Bottle
4947.97 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(pyrid-3-ylmethyl)piperidine-4-carboxylic acid dihydrochloride, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.