Learn More
1-Pyrrolidino-1-cyclohexene, 95%, Thermo Scientific Chemicals
697.01 SEK - 1940.74 SEK
Chemical Identifiers
CAS | 1125-99-1 |
---|---|
Molecular Formula | C10H17N |
Molecular Weight (g/mol) | 151.25 |
MDL Number | MFCD00003163 |
InChI Key | KTZNVZJECQAMBV-UHFFFAOYSA-N |
Synonym | 1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine |
PubChem CID | 70768 |
IUPAC Name | 1-(cyclohexen-1-yl)pyrrolidine |
SMILES | C1CCC(=CC1)N2CCCC2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10552701
|
Thermo Scientific Acros
334830250 |
25 g | Glass bottle |
697.01 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307581
|
Thermo Scientific Acros
334831000 |
100 g | Glass bottle |
1940.74 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1125-99-1 | |
151.25 | |
KTZNVZJECQAMBV-UHFFFAOYSA-N | |
70768 | |
C1CCC(=CC1)N2CCCC2 |
C10H17N | |
MFCD00003163 | |
1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine | |
1-(cyclohexen-1-yl)pyrrolidine |
Specifications
1125-99-1 | |
100.0 | |
0.9400g/mL | |
66°C | |
94% min. (GC) | |
1.5210 to 1.523 | |
0.94 | |
KTZNVZJECQAMBV-UHFFFAOYSA-N | |
1-(cyclohexen-1-yl)pyrrolidine | |
70768 | |
95% | |
1-Pyrrolidino-1-cyclohexene, 95% |
94.0 | |
Yellow | |
114°C to 115°C (15.0 mmHg) | |
Authentic | |
C10H17N | |
MFCD00003163 | |
1-pyrrolidino-1-cyclohexene, 1-pyrrolidinocyclohexene, 1-cyclohex-1-en-1-yl pyrrolidine, pyrrolidine, 1-1-cyclohexen-1-yl, 1-1-cyclohexen-1-yl pyrrolidine, 1-1-pyrrolidinyl cyclohexene, cyclohexanone pyrrolidine enamine, 1-pyrrolidinyl-1-cyclohexene, n-1-cyclohexenyl pyrrolidine, 1-1-cyclohexenyl pyrrolidine | |
C1CCC(=CC1)N2CCCC2 | |
151.25 | |
151.25 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 214-414-6
missing translation for 'tsca' : TSCA
- 1-Pyrrolidino-1-cyclohexene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only