Learn More
Axygen™ 1250μL Universal Pipette Tips
GREENER_CHOICE
Matrix compatible tips in clear color
Brand:  Axygen™ MTX-1250-C
 View more versions of this product
Product Code. 11386324
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Matrix-style tips fits Matrix Impact™ and Impact2™ brand pipettors
  • Maxymum Recovery™ tips allows complete retrieval of precious samples
  • Optimized tip bevel reduces sample hang-up and improves pipetting accuracy
  • Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
  • Volume range: 15 to 1250μL
  • Available in bulk or racked formats
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1250 μL
Clear
RNase, DNase-free
Matrix™ Impact™ Pipette
Non-sterile
Autoclavable
Bulk
Polypropylene