missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products 1422A DuPont™ Tyvek™ Autoclave Sterilization Wrapper
BIOPHARMAGREENER_CHOICE

Autoclavable Wrapper provides an excellent bacterial and microbial barrier for superior performance post steam sterilization.

Brand:  Keystone Cleanroom Products T2ASHEET4545

 View more versions of this product

Product Code. 17728547

  • 18355.00 SEK / Pack of 125
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Tyvek™ Autoclavable Wrappers are breathable while providing an excellent bacterial and microbial barrier for superior performance post steam sterilization.

  • Lint-free (Helmke Drum) means no airborne particulates are generated during its use as sterilization packaging
  • Clean and consistently cut using an automated CAD system per ISO 8 Requirements
  • Packaged in an ISO 6 Controlled Clean Room
  • Designed for Steam Sterilization
  • Hydrophobic
  • Resists tearing and punctures
Specifications

Specifications

Sterilization Wrap
COC
Single-use
Pharmaceutical Production Equipment
Non-sterile
Yes
Steam, EtO, Gamma, VHP
<125°C
45 in.
45 in.
Autoclavable
White
Steam Sterilization
HDPE
5.3 mm
125
<250°F
45 x 45 in.
114.3 cm
114.3 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.