Learn More
Chem Lab 1LT Sulfuric acid 18 mol/l (1765.44 g H2SO4 / lH2O = 36 N (±0.072/20°C))
Brand: Chem Lab CL05.2628.1000
1LT Sulfuric acid 18 mol/l (1765.44 g H2SO4 / lH2O = 36 N (±0.072/20°C))
This item is not returnable.
View return policy
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sulphuric acid 18 mol/l
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sulphuric acid
Product Content Correction
Product Title
Chem Lab 1LT Sulfuric acid 18 mol/l (1765.44 g H2SO4 / lH2O = 36 N (±0.072/20°C)) >
Spot an opportunity for improvement?Share a Content Correction