Learn More
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 99%, 99% ee, Thermo Scientific Chemicals
715.11 SEK - 1802.04 SEK
Chemical Identifiers
CAS | 35132-20-8 |
---|---|
Molecular Formula | C14H18N2 |
Molecular Weight (g/mol) | 214.31 |
MDL Number | MFCD00082769 |
InChI Key | PONXTPCRRASWKW-ZIAGYGMSSA-P |
Synonym | 1r,2r-+-1,2-diphenylethylenediamine, 1r,2r-1,2-diphenylethane-1,2-diamine, 1r,2r-rel-1,2-diphenylethane-1,2-diamine, 1r,2r-1,2-diphenylethylenediamine, 1r,2r-+-1,2-diamino-1,2-diphenylethane, 1r,2r-1,2-diphenyl-1,2-ethanediamine, 1r,2r-+-1,2-diphenyl-1,2-ethanediamine, +-1,2-diphenylethylenediamine, +-stilbenediamine, r,r-dpen |
PubChem CID | 2724998 |
IUPAC Name | (1R,2R)-1,2-diphenylethane-1,2-diamine |
SMILES | [NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10043550
|
Thermo Scientific Acros
294682500 |
250 mg | Glass bottle |
715.11 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10209173
|
Thermo Scientific Acros
294680010 |
1 g | Glass bottle |
1802.04 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
35132-20-8 | |
214.31 | |
PONXTPCRRASWKW-ZIAGYGMSSA-P | |
2724998 | |
[NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1 |
C14H18N2 | |
MFCD00082769 | |
1r,2r-+-1,2-diphenylethylenediamine, 1r,2r-1,2-diphenylethane-1,2-diamine, 1r,2r-rel-1,2-diphenylethane-1,2-diamine, 1r,2r-1,2-diphenylethylenediamine, 1r,2r-+-1,2-diamino-1,2-diphenylethane, 1r,2r-1,2-diphenyl-1,2-ethanediamine, 1r,2r-+-1,2-diphenyl-1,2-ethanediamine, +-1,2-diphenylethylenediamine, +-stilbenediamine, r,r-dpen | |
(1R,2R)-1,2-diphenylethane-1,2-diamine |
Specifications
35132-20-8 | |
100.0 | |
White to Yellow | |
Authentic | |
C14H18N2 | |
MFCD00082769 | |
+ 104.00 | |
PONXTPCRRASWKW-ZIAGYGMSSA-P | |
(1R,2R)-1,2-diphenylethane-1,2-diamine | |
2724998 | |
99%,99% ee | |
(1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine, 99% (99% e.e.) |
98.5 | |
81.0°C to 84.0°C | |
98.5% min. | |
98.5% min. (GC) | |
(C6H5CH(NH2)-)2 | |
+ 104.00 (25.00°C c=1.1,MeOH) | |
1r,2r-+-1,2-diphenylethylenediamine, 1r,2r-1,2-diphenylethane-1,2-diamine, 1r,2r-rel-1,2-diphenylethane-1,2-diamine, 1r,2r-1,2-diphenylethylenediamine, 1r,2r-+-1,2-diamino-1,2-diphenylethane, 1r,2r-1,2-diphenyl-1,2-ethanediamine, 1r,2r-+-1,2-diphenyl-1,2-ethanediamine, +-1,2-diphenylethylenediamine, +-stilbenediamine, r,r-dpen | |
[NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1 | |
214.31 | |
212.29 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'tsca' : TSCA
- (1R,2R)-(+)-1,2-Diphenyl-1,2-ethanediamine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.