Learn More
2-(1,4-Diazepan-1-yl)nicotinonitrile, ≥95%, Thermo Scientific™
529.98 SEK - 1407.22 SEK
Chemical Identifiers
CAS | 352018-97-4 |
---|---|
Molecular Formula | C11H14N4 |
Molecular Weight (g/mol) | 202.261 |
MDL Number | MFCD03086210 |
InChI Key | KQOHMUWBQXSUAK-UHFFFAOYSA-N |
Synonym | 2-1,4-diazepan-1-yl nicotinonitrile, 2-1,4-diazepan-1-yl pyridine-3-carbonitrile, 3-pyridinecarbonitrile,2-hexahydro-1h-1,4-diazepin-1-yl, 2-homopiperazin-1-yl nicotinonitrile, 2-1,4 diazepan-1-yl-nicotinonitrile, 2-1,4-diazaperhydroepinyl pyridine-3-carbonitrile |
PubChem CID | 2779669 |
IUPAC Name | 2-(1,4-diazepan-1-yl)pyridine-3-carbonitrile |
SMILES | C1CNCCN(C1)C2=C(C=CC=N2)C#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10671873
|
Thermo Scientific Maybridge
MO00102CB |
250 mg | Amber Glass Bottle |
529.98 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10561112
|
Thermo Scientific Maybridge
MO00102DA |
1 g | Amber Glass Bottle |
1407.22 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
352018-97-4 | |
202.261 | |
KQOHMUWBQXSUAK-UHFFFAOYSA-N | |
2779669 | |
C1CNCCN(C1)C2=C(C=CC=N2)C#N |
C11H14N4 | |
MFCD03086210 | |
2-1,4-diazepan-1-yl nicotinonitrile, 2-1,4-diazepan-1-yl pyridine-3-carbonitrile, 3-pyridinecarbonitrile,2-hexahydro-1h-1,4-diazepin-1-yl, 2-homopiperazin-1-yl nicotinonitrile, 2-1,4 diazepan-1-yl-nicotinonitrile, 2-1,4-diazaperhydroepinyl pyridine-3-carbonitrile | |
2-(1,4-diazepan-1-yl)pyridine-3-carbonitrile |
Specifications
352018-97-4 | |
95% | |
MFCD03086210 | |
KQOHMUWBQXSUAK-UHFFFAOYSA-N | |
2-(1,4-diazepan-1-yl)pyridine-3-carbonitrile | |
2779669 | |
95% |
95.0 | |
C11H14N4 | |
2-1,4-diazepan-1-yl nicotinonitrile, 2-1,4-diazepan-1-yl pyridine-3-carbonitrile, 3-pyridinecarbonitrile,2-hexahydro-1h-1,4-diazepin-1-yl, 2-homopiperazin-1-yl nicotinonitrile, 2-1,4 diazepan-1-yl-nicotinonitrile, 2-1,4-diazaperhydroepinyl pyridine-3-carbonitrile | |
C1CNCCN(C1)C2=C(C=CC=N2)C#N | |
202.261 | |
202.26 | |
2-(1, 4-Diazepan-1-yl)nicotinonitrile |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.