missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl trifluoromethanesulfonate, 95%, Thermo Scientific Chemicals

1480.00 SEK - 5700.00 SEK

Chemical Identifiers

CAS 6226-25-1
Molecular Formula C3H2F6O3S
Molecular Weight (g/mol) 232.096
MDL Number MFCD00671579
InChI Key RTMMSCJWQYWMNK-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
PubChem CID 80366
IUPAC Name 2,2,2-trifluoroethyl trifluoromethanesulfonate
SMILES C(C(F)(F)F)OS(=O)(=O)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15496636
SDS View Documents
Thermo Scientific Alfa Aesar
H61603.06
5 g
1480.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
15406646
SDS View Documents
Thermo Scientific Alfa Aesar
H61603.14
25 g
5700.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,2,2-Trifluoroethyl trifluoromethanesulfonate is a powerful trifluoroethylating agent which is useful for synthesis of fluorinated amino acids. It is used as a reagent in thenantioselective preparation of cyclic N-aryl hydroxamic acids via phase-transfer catalyzed alkylation of nitrobenzyl bromides to give nirophenylalanines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroethyl trifluoromethanesulfonate is a powerful trifluoroethylating agent which is useful for synthesis of fluorinated amino acids. It is used as a reagent in thenantioselective preparation of cyclic N-aryl hydroxamic acids via phase-transfer catalyzed alkylation of nitrobenzyl bromides to give nirophenylalanines.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'rtecsNumber' : PB2775000

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2,2-Trifluoroethyl trifluoromethanesulfonate, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.