missing translation for 'onlineSavingsMsg'
Learn More
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol, 97%, Thermo Scientific Chemicals Product Code.: 11309235

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol, 97%, Thermo Scientific Chemicals

Product Code. 11309235
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11309235

Brand: Thermo Scientific Alfa Aesar L16560.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-74-8
Molecular Formula C6H6F8O2
Molecular Weight (g/mol) 262.099
MDL Number MFCD00069087
InChI Key NHEKBXPLFJSSBZ-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,5,5-octafluoro-1,6-hexanediol, 1,6-dihydroxy-2,2,3,3,4,4,5,5-octafluorohexane, 1,6-hexanediol, 2,2,3,3,4,4,5,5-octafluoro, 2,2,3,3,4,4,5,5-octafluorohexan-1,6-diol, acmc-1code, nhekbxplfjssbz-uhfffaoysa, 2,2,3,3,4,4,5,5octafluoro-1,6-hexanediol
PubChem CID 136181
IUPAC Name 2,2,3,3,4,4,5,5-octafluorohexane-1,6-diol
SMILES C(C(C(C(C(CO)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 67°C to 70°C
Boiling Point 100°C (3 mmHg)
Quantity 25 g
Beilstein 1784039
Solubility Information Soluble in water.
Formula Weight 262.1
Percent Purity 97%
Chemical Name or Material 2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10207499 Certificate TypeCertificate of Analysis Date08/08/2023 Catalog NumberL16560.14,L16560.06
Lot NumberK18X023 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16560.14,L16560.06
Lot Number10175815 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL16560.14,L16560.06
Lot Number10234680 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL16560.14,L16560.06
Lot Number10182185 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL16560.14,L16560.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.