missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Dichlorophenoxy)thioacetamide, 97%, Thermo Scientific™

Product Code. 15433497
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2302-32-1
Molecular Formula C8H7Cl2NOS
Molecular Weight (g/mol) 236.11
MDL Number MFCD00172128
InChI Key AGDXYFYNDRKKSF-UHFFFAOYSA-N
Synonym 2-2,4-dichlorophenoxy ethanethioamide, ethanethioamide,2-2,4-dichlorophenoxy, acmc-20aorc, dichlorophenoxyethanethioamide, 2-2,4-dichlorophenoxy thioacetamide, 2-2,4-dichlorophenoxy-thioacetamide, 1-amino-2-2,4-dichlorophenoxy ethane-1-thione
PubChem CID 3262582
IUPAC Name 2-(2,4-dichlorophenoxy)ethanethioamide
SMILES C1=CC(=C(C=C1Cl)Cl)OCC(=S)N

Specifications

Melting Point 136°C to 138°C
Quantity 250 mg
Formula Weight 236.11
Percent Purity 97%
Chemical Name or Material 2-(2,4-Dichlorophenoxy)thioacetamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'