missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid, 97%, Thermo Scientific™
813.49 SEK - 3136.45 SEK
Chemical Identifiers
CAS | 656-46-2 |
---|---|
Molecular Formula | C8H4F2O4 |
Molecular Weight (g/mol) | 202.12 |
InChI Key | VJLDRFCNFNQTTH-UHFFFAOYSA-N |
Synonym | 2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid, 2,2-difluorobenzodioxole-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid, 3,4-difluoromethylenedioxy benzoic acid, acmc-209nrl, 5-carboxy-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo-1,3dioxole-5-carboxylic acid, 2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acidShow More |
PubChem CID | 608772 |
IUPAC Name | 2,2-difluoro-1,3-benzodioxole-5-carboxylic acid |
SMILES | C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
15189323
|
Thermo Scientific Acros
449140010 |
1 g | Glass bottle | N/A | ||||||
15199323
|
Thermo Scientific Acros
449140050 |
5 g | Glass bottle |
3136.45 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
656-46-2 | |
202.12 | |
2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid, 2,2-difluorobenzodioxole-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid, 3,4-difluoromethylenedioxy benzoic acid, acmc-209nrl, 5-carboxy-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo-1,3dioxole-5-carboxylic acid, 2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acidShow More | |
2,2-difluoro-1,3-benzodioxole-5-carboxylic acid |
C8H4F2O4 | |
VJLDRFCNFNQTTH-UHFFFAOYSA-N | |
608772 | |
C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F |
Specifications
656-46-2 | |
96% min. (HPLC) | |
2,2-difluorobenzo d 1,3 dioxole-5-carboxylic acid, 2,2-difluoro-2h-1,3-benzodioxole-5-carboxylic acid, 2,2-difluorobenzodioxole-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxole-5-carboxylic acid, 3,4-difluoromethylenedioxy benzoic acid, acmc-209nrl, 5-carboxy-2,2-difluoro-1,3-benzodioxole, 2,2-difluorobenzo-1,3dioxole-5-carboxylic acid, 2,2-difluorobenzo 1,3 dioxol-5-carboxylic acid, 2,2-difluoro-benzo 1,3 dioxol-5-carboxylic acid | |
C1=CC2=C(C=C1C(=O)O)OC(O2)(F)F | |
202.12 | |
202.12 | |
2, 2-Difluoro-1, 3-benzodioxole-5-carboxylic acid |
153°C to 157°C | |
C8H4F2O4 | |
VJLDRFCNFNQTTH-UHFFFAOYSA-N | |
2,2-difluoro-1,3-benzodioxole-5-carboxylic acid | |
608772 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid, 97%, Thermo Scientific™