Learn More
2,3-Butanedione monoxime, 99%, Thermo Scientific Chemicals
Description
2,3-Butanedione monoxime is a reversible inhibitor of myosin ATPase. It is also used as a reagent for colorimetric determination of urea.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,3-Butanedione monoxime is a reversible inhibitor of myosin ATPase. It is also used as a reagent for colorimetric determination of urea.
Solubility
Soluble in water (50g/L at 20°C)
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 74°C to 77°C |
Boiling Point | 185°C to 186°C |
Quantity | 500 g |
Beilstein | 605582 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water (50g/L at 20°C) |
Formula Weight | 101.11 |
Percent Purity | 99% |
Chemical Name or Material | 2,3-Butanedione monoxime |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10249478 | Certificate of Analysis | 07/02/2024 | A14339.0E,A14339.36,A14339.22 |
10244102 | Certificate of Analysis | 19/10/2022 | A14339.0E,A14339.36,A14339.22 |
10238824 | Certificate of Analysis | 12/04/2022 | A14339.0E,A14339.36,A14339.22 |
10142631 | Certificate of Analysis | 22/11/2021 | A14339.0E,A14339.36,A14339.22 |
10211283 | Certificate of Analysis | 22/11/2021 | A14339.0E,A14339.36,A14339.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.