missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diaminopyridine, 98%, Thermo Scientific Chemicals

Product Code. 11487556
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.

Solubility
Soluble in water (100 g/L).

Notes
It is sensitive to air. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-58-4
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006319
InChI Key ZZYXNRREDYWPLN-UHFFFAOYSA-N
Synonym 2,3-diaminopyridine, 2,3-pyridinediamine, pyridine, 2,3-diamino, pyridine-2,3-diyldiamine, unii-cbx394737h, diaminopyridine, 2,3-diamino-pyridine, pyridinediamine, pyridine,3-diamino, pubchem1296
PubChem CID 9956
IUPAC Name pyridine-2,3-diamine
SMILES NC1=CC=CN=C1N

Specifications

Melting Point 110°C to 114°C
Boiling Point 195°C (55 mmHg)
Flash Point 205°C (401°F)
Quantity 5 g
UN Number UN2671
Beilstein 109869
Sensitivity Air Sensitive
Solubility Information Soluble in water (100g/L).
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2,3-Diaminopyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Diaminopyridine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.