missing translation for 'onlineSavingsMsg'
Learn More
2,3-Dichloro-6-fluorobenzyl bromide, 97%, Thermo Scientific™ Product Code.: 11336909

2,3-Dichloro-6-fluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 11336909
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11336909

Brand: Thermo Scientific Alfa Aesar H26800.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886497-51-4
Molecular Formula C7H4BrCl2F
Molecular Weight (g/mol) 257.911
MDL Number MFCD04115873
InChI Key RXHTVFRUZNHYSD-UHFFFAOYSA-N
Synonym 2,3-dichloro-6-fluorobenzyl bromide, 2-bromomethyl-3,4-dichloro-1-fluorobenzene, 2-bromomethyl-3,4-dichloro-1-fluoro-benzene, 1,2-dichloro-4-fluoro-3-bromomethyl benzene, 3-bromomethyl-1,2-dichloro-4-fluorobenzene, benzene,2-bromomethyl-3,4-dichloro-1-fluoroShow MoreShow Less
PubChem CID 3824079
IUPAC Name 3-(bromomethyl)-1,2-dichloro-4-fluorobenzene
SMILES C1=CC(=C(C(=C1F)CBr)Cl)Cl

Specifications

Quantity 1 g
UN Number UN3265
Formula Weight 257.92
Percent Purity 97%
Chemical Name or Material 2,3-Dichloro-6-fluorobenzyl bromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10150749 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10151439 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10152580 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10141230 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3-Dichloro-6-fluorobenzyl bromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.