Learn More
2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate, 95%, Thermo Scientific Chemicals
865.00 SEK - 2670.00 SEK
Chemical Identifiers
CAS | 207233-91-8 |
---|---|
Molecular Formula | C3H7O3S3 |
Molecular Weight (g/mol) | 187.27 |
MDL Number | MFCD00149543 |
InChI Key | JLVSRWOIZZXQAD-GSVOUGTGSA-M |
Synonym | sodium 2,3-dimercaptopropanesulfonate monohydrate, unithiol, dl-2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropane-1-sulfonate hydrate, 2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropanesulfonate hydrate, dmps, sodium 2,3-disulfanylpropane-1-sulfonate hydrate, unitiol monohydrate |
PubChem CID | 23676755 |
SMILES | [O-]S(=O)(=O)C[C@H](S)CS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442776
|
Thermo Scientific Alfa Aesar
H56578.MD |
250 mg |
865.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15432776
|
Thermo Scientific Alfa Aesar
H56578.03 |
1 g |
2670.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium 2,3-dimercaptopropanesulfonate monohydrate has been used in a study to assess its use as an antidote.
Solubility
Soluble in Water: 100 mg/ml
Notes
Store at 4°C. Incompatible with oxidizing agents. Keep container tightly closed.
Chemical Identifiers
207233-91-8 | |
187.27 | |
JLVSRWOIZZXQAD-GSVOUGTGSA-M | |
23676755 |
C3H7O3S3 | |
MFCD00149543 | |
sodium 2,3-dimercaptopropanesulfonate monohydrate, unithiol, dl-2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropane-1-sulfonate hydrate, 2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropanesulfonate hydrate, dmps, sodium 2,3-disulfanylpropane-1-sulfonate hydrate, unitiol monohydrate | |
[O-]S(=O)(=O)C[C@H](S)CS |
Specifications
207233-91-8 | |
C3H7O3S3 | |
3734863 | |
sodium 2,3-dimercaptopropanesulfonate monohydrate, unithiol, dl-2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropane-1-sulfonate hydrate, 2,3-dimercapto-1-propanesulfonic acid sodium salt monohydrate, sodium 2,3-dimercaptopropanesulfonate hydrate, dmps, sodium 2,3-disulfanylpropane-1-sulfonate hydrate, unitiol monohydrate | |
JLVSRWOIZZXQAD-GSVOUGTGSA-M | |
(2R)-2,3-disulfanylpropane-1-sulfonate | |
23676755 | |
98% |
∽229°C (decomposition) | |
MFCD00149543 | |
14,3210 | |
Soluble in Water: 100mg/ml | |
[O-]S(=O)(=O)C[C@H](S)CS | |
187.27 | |
228.30 (210.28 Anhydrous) | |
2,3-Dimercapto-1-propanesulfonic acid sodium salt monohydrate |
Safety and Handling
missing translation for 'einecsNumber' : 223-796-3
missing translation for 'rtecsNumber' : TZ6420000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.