missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimercapto-1-propanol, Honeywell Fluka™
Click to view available options
Quantity:
5 mL
25 mL
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Density | 1.239g/mL at 25°C |
Flash Point | 112°C (233.6°F) |
Packaging | Glass Bottle |
Linear Formula | HOCH2CH(SH)CH2SH |
Quantity | 25 mL |
UN Number | UN2810 |
Beilstein | 1732058 |
Formula Weight | 124.23g/mol |
Percent Purity | ≥98% |
Chemical Name or Material | 2,3-Dimercapto-1-propanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'