missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorophenylboronic acid, 97%, Thermo Scientific™

1608.15 SEK - 4226.00 SEK

Chemical Identifiers

CAS 247564-72-3
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.90
MDL Number MFCD01863165
InChI Key KCHHKNCSISEAAE-UHFFFAOYSA-N
Synonym 2,4,5-trifluorophenyl boronic acid, 2,4,5-trifluorobenzeneboronic acid, boronic acid, b-2,4,5-trifluorophenyl, boronic acid, 2,4,5-trifluorophenyl, 2,4,5-trifluorophenylboronicacid, pubchem1787, acmc-1cd74Show MoreShow Less
PubChem CID 2783133
IUPAC Name (2,4,5-trifluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(F)C=C1F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11925791
View Documents
Thermo Scientific Acros
393800050
5 g
1608.15 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11935791
View Documents
Thermo Scientific Acros
393800250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4,5-Trifluorophenylboronic acid, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.