missing translation for 'onlineSavingsMsg'
Learn More
2,4,5-Triphenylimidazole, 97%, Thermo Scientific Chemicals Product Code.: 11433527

2,4,5-Triphenylimidazole, 97%, Thermo Scientific Chemicals

Product Code. 11433527
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11433527

Brand: Thermo Scientific Alfa Aesar A17291.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 484-47-9
Molecular Formula C21H16N2
Molecular Weight (g/mol) 296.373
MDL Number MFCD00005187
InChI Key RNIPJYFZGXJSDD-UHFFFAOYSA-N
Synonym 2,4,5-triphenylimidazole, lophine, lophin, 1h-imidazole, 2,4,5-triphenyl, 2,4,5-triphenyl-1,3-imidazole, unii-q6k46g80zd, imidazole, 2,4,5-triphenyl, chembl34173, 2,5-triphenylimidazole, imidazole,4,5-triphenylShow MoreShow Less
PubChem CID 10232
IUPAC Name 2,4,5-triphenyl-1H-imidazole
SMILES C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 274°C to 278°C
Quantity 50 g
Beilstein 236652
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 296.38
Percent Purity 97%
Chemical Name or Material 2,4,5-Triphenylimidazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX02J008 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA17291.30,A17291.09,A17291.18
Lot NumberX21I001 Certificate TypeCertificate of Analysis Date28/10/2022 Catalog NumberA17291.30,A17291.09,A17291.18
Lot Number10235918 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17291.30,A17291.09,A17291.18
Lot NumberJ19U008 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17291.30,A17291.09,A17291.18
Lot NumberR21A020 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17291.30,A17291.09,A17291.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4,5-Triphenylimidazole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.