missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Product Code. 11469530
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61040-78-6
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00060356
InChI Key CFXXBVNHYJQNKS-UHFFFAOYSA-N
PubChem CID 602515
IUPAC Name (2,4,6-trimethoxyphenyl)methanol
SMILES COC1=CC(=C(C(=C1)OC)CO)OC

Specifications

Melting Point 60°C
Quantity 1 g
Synonym 2,4,6-trimethoxyphenyl methanol, 2,4,6-trimethoxybenzyl alcohol, 2,4,6-trimethoxyphenyl methan-1-ol, 2,4,6-trimethoxy-benzyl alcohol, benzenemethanol,2,4,6-trimethoxy, benzenemethanol, 2,4,6-trimethoxy, 2,4,6-trimethoxy-phenyl-methanol, 2,4,6-trimethoxyphenyl methanol #, 3-amino-n-1,1-dimethylethoxy carbonyl-l-alanine methyl ester
Formula Weight 198.22
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethoxybenzyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'