missing translation for 'onlineSavingsMsg'
Learn More
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester, 99%, Thermo Scientific™ Product Code.: 11314006

2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester, 99%, Thermo Scientific™

Product Code. 11314006
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11314006

Brand: Thermo Scientific Alfa Aesar L17232.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214360-78-8
Molecular Formula C14H21BO2
Molecular Weight (g/mol) 232.13
MDL Number MFCD01318178
InChI Key ITLDRNGUSKUMRG-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-2,4,6-trimethylphenyl-1,3,2-dioxaborinane, 2-mesityl-5,5-dimethyl-1,3,2-dioxaborinane, 2,4,6-trimethylbenzeneboronic acid neopentyl glycol cyclic ester, mesitylboronic acid 2,2-dimethyltrimethylene ester, 2,4,6-trimethyl-benzeneboronic acid neopentyl glycol cyclic ester, 2,4,6-trimethylphenylboronic acid neopentyl glycol ester, mesityleneboronic acid 2,2-dimethyltrimethylene cyclic esterShow MoreShow Less
PubChem CID 11436188
IUPAC Name 5,5-dimethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborinane
SMILES B1(OCC(CO1)(C)C)C2=C(C=C(C=C2C)C)C

Specifications

Melting Point 46°C to 48°C
Quantity 5 g
Beilstein 8689636
Formula Weight 232.13
Percent Purity 99%
Chemical Name or Material 2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.