Learn More
2-(4-Bromophenoxy)tetrahydropyran, 98%, Thermo Scientific Chemicals
659.00 SEK - 2000.00 SEK
Chemical Identifiers
CAS | 36603-49-3 |
---|---|
Molecular Formula | C11H13BrO2 |
Molecular Weight (g/mol) | 257.127 |
MDL Number | MFCD00091551 |
InChI Key | MXDQGXMBJCGRCB-UHFFFAOYSA-N |
Synonym | 2-4-bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy tetrahydropyran, 2-4-bromophenoxy oxane, 2h-pyran, 2-4-bromophenoxy tetrahydro, 2-4'-bromophenoxy-tetrahydropyran, 4-bromophenyl thp ether, acmc-209inn, bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy-tetrahydropyran |
PubChem CID | 4646436 |
IUPAC Name | 2-(4-bromophenoxy)oxane |
SMILES | C1CCOC(C1)OC2=CC=C(C=C2)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11485660
|
Thermo Scientific Alfa Aesar
H25914.09 |
10 g |
659.00 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495660
|
Thermo Scientific Alfa Aesar
H25914.18 |
50 g |
2000.00 SEK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-(4-Bromophenoxy)tetrahydropyran can be used to produce 4-bromo-phenol at temperature of 20°C. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(4-Bromophenoxy)tetrahydropyran can be used to produce 4-bromo-phenol at temperature of 20°C. It is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Chemical Identifiers
36603-49-3 | |
257.127 | |
MXDQGXMBJCGRCB-UHFFFAOYSA-N | |
4646436 | |
C1CCOC(C1)OC2=CC=C(C=C2)Br |
C11H13BrO2 | |
MFCD00091551 | |
2-4-bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy tetrahydropyran, 2-4-bromophenoxy oxane, 2h-pyran, 2-4-bromophenoxy tetrahydro, 2-4'-bromophenoxy-tetrahydropyran, 4-bromophenyl thp ether, acmc-209inn, bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy-tetrahydropyran | |
2-(4-bromophenoxy)oxane |
Specifications
36603-49-3 | |
C11H13BrO2 | |
2-4-bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy tetrahydropyran, 2-4-bromophenoxy oxane, 2h-pyran, 2-4-bromophenoxy tetrahydro, 2-4'-bromophenoxy-tetrahydropyran, 4-bromophenyl thp ether, acmc-209inn, bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy-tetrahydropyran | |
MXDQGXMBJCGRCB-UHFFFAOYSA-N | |
2-(4-bromophenoxy)oxane | |
4646436 | |
98% |
55°C to 58°C | |
MFCD00091551 | |
Slightly soluble in water. | |
C1CCOC(C1)OC2=CC=C(C=C2)Br | |
257.127 | |
257.13 | |
2-(4-Bromophenoxy)tetrahydropyran |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures