missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)nicotinic acid, 98%, Thermo Scientific™

415.00 SEK - 1207.04 SEK

Chemical Identifiers

CAS 51362-37-9
Molecular Formula C12H8ClNO3
Molecular Weight (g/mol) 249.65
MDL Number MFCD00052116
InChI Key FVAZJGJEENAKRJ-UHFFFAOYSA-N
Synonym 2-4-chlorophenoxy nicotinic acid, 2-4-chlorophenoxy pyridine-3-carboxylic acid, 2-4-chloro-phenoxy-nicotinic acid, 3-pyridinecarboxylic acid, 2-4-chlorophenoxy, acmc-1avkg, maybridge1_001700, 3-pyridinecarboxylicacid, 2-4-chlorophenoxy
PubChem CID 609454
IUPAC Name 2-(4-chlorophenoxy)pyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)OC2=CC=C(C=C2)Cl)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11496927
View Documents
Thermo Scientific Alfa Aesar
A14359.03
1 g
415.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11406937
View Documents
Thermo Scientific Alfa Aesar
A14359.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

51362-37-9
249.65
FVAZJGJEENAKRJ-UHFFFAOYSA-N
609454
C1=CC(=C(N=C1)OC2=CC=C(C=C2)Cl)C(=O)O
C12H8ClNO3
MFCD00052116
2-4-chlorophenoxy nicotinic acid, 2-4-chlorophenoxy pyridine-3-carboxylic acid, 2-4-chloro-phenoxy-nicotinic acid, 3-pyridinecarboxylic acid, 2-4-chlorophenoxy, acmc-1avkg, maybridge1_001700, 3-pyridinecarboxylicacid, 2-4-chlorophenoxy
2-(4-chlorophenoxy)pyridine-3-carboxylic acid

Specifications

51362-37-9
C12H8ClNO3
2-4-chlorophenoxy nicotinic acid, 2-4-chlorophenoxy pyridine-3-carboxylic acid, 2-4-chloro-phenoxy-nicotinic acid, 3-pyridinecarboxylic acid, 2-4-chlorophenoxy, acmc-1avkg, maybridge1_001700, 3-pyridinecarboxylicacid, 2-4-chlorophenoxy
C1=CC(=C(N=C1)OC2=CC=C(C=C2)Cl)C(=O)O
249.65
249.64
2-(4-Chlorophenoxy)nicotinic acid
157°C to 159°C
MFCD00052116
FVAZJGJEENAKRJ-UHFFFAOYSA-N
2-(4-chlorophenoxy)pyridine-3-carboxylic acid
609454
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications