Learn More
2,4-Pentanedione dioxime, 98+%, Thermo Scientific™
480.00 SEK - 1468.11 SEK
Chemical Identifiers
CAS | 2157-56-4 |
---|---|
Molecular Formula | C5H10N2O2 |
Molecular Weight (g/mol) | 130.15 |
MDL Number | MFCD00013932 |
InChI Key | WBRYLZHYOFBTPD-YDFGWWAZSA-N |
Synonym | acetylacetone dioxime, 2,4-pentanedione dioxime, 2,4-pentanedione, dioxime, 2,4-pentanedioxime, 2,4-pentanedione oxime, 2,4-pentanedione,2,4-dioxime, 2,4-pentanedione, 2,4-dioxime, pentane-2,4-dione oxime, 2e,4e-2,4-pentanedione dioxime #, ne-n-4e-4-hydroxyiminopentan-2-ylidene hydroxylamine |
PubChem CID | 9573501 |
SMILES | C\C(C\C(C)=N\O)=N/O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429903
|
Thermo Scientific Alfa Aesar
A13003.06 |
5 g |
480.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439903
|
Thermo Scientific Alfa Aesar
A13003.14 |
25 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2157-56-4 | |
130.15 | |
WBRYLZHYOFBTPD-YDFGWWAZSA-N | |
9573501 |
C5H10N2O2 | |
MFCD00013932 | |
acetylacetone dioxime, 2,4-pentanedione dioxime, 2,4-pentanedione, dioxime, 2,4-pentanedioxime, 2,4-pentanedione oxime, 2,4-pentanedione,2,4-dioxime, 2,4-pentanedione, 2,4-dioxime, pentane-2,4-dione oxime, 2e,4e-2,4-pentanedione dioxime #, ne-n-4e-4-hydroxyiminopentan-2-ylidene hydroxylamine | |
C\C(C\C(C)=N\O)=N/O |
Specifications
2157-56-4 | |
C5H10N2O2 | |
4372703 | |
WBRYLZHYOFBTPD-YDFGWWAZSA-N | |
(NE)-N-[(4E)-4-hydroxyiminopentan-2-ylidene]hydroxylamine | |
9573501 | |
≥98% |
147°C to 151°C | |
MFCD00013932 | |
acetylacetone dioxime, 2,4-pentanedione dioxime, 2,4-pentanedione, dioxime, 2,4-pentanedioxime, 2,4-pentanedione oxime, 2,4-pentanedione,2,4-dioxime, 2,4-pentanedione, 2,4-dioxime, pentane-2,4-dione oxime, 2e,4e-2,4-pentanedione dioxime #, ne-n-4e-4-hydroxyiminopentan-2-ylidene hydroxylamine | |
C\C(C\C(C)=N\O)=N/O | |
130.15 | |
130.15 | |
2,4-Pentanedione dioxime |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 218-472-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures