missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diamino-3-iodopyridine, 95%, Thermo Scientific™

Product Code. 15464117
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1125410-09-4
Molecular Formula C5H6IN3
MDL Number MFCD11844805
Quantity 250 mg
Sensitivity Light sensitive
Synonym 2,5-diamino-3-iodopyridine, 2,5-pyridinediamine, 3-iodo, 3-iodo-pyridine-2,5-diamine
InChI Key SECWUDQFVZPDOS-UHFFFAOYSA-N
SMILES C1=C(C=NC(=C1I)N)N
IUPAC Name 3-iodopyridine-2,5-diamine
Molecular Weight (g/mol) 235.028
PubChem CID 45588197
Formula Weight 235.03
Percent Purity 95%
Chemical Name or Material 2,5-Diamino-3-iodopyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'