missing translation for 'onlineSavingsMsg'
Learn More
2,6-Dibromopyridine, 98%, Thermo Scientific Chemicals Product Code.: 11448356

2,6-Dibromopyridine, 98%, Thermo Scientific Chemicals

Product Code. 11448356
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11448356

Brand: Thermo Scientific Alfa Aesar A15397.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromopyridine is used as a tridentate chelating ligand and in the formation of macrocycles containing the terpyridine moiety. It is also used to produce 6-bromo-2-methoxypyridine.

Solubility
Soluble in hot methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-05-1
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.894
MDL Number MFCD00006223
InChI Key FEYDZHNIIMENOB-UHFFFAOYSA-N
Synonym pyridine, 2,6-dibromo, 2,6-dibromo-pyridine, 2,6-dibromo pyridine, 2,6dibromopyridine, 2,6-dibrompyridine, 2,6 dibromopyridine, 2.6-dibromopyridine, pubchem3204, 2, 6-dibromopyridine, acmc-209n5v
PubChem CID 12274
IUPAC Name 2,6-dibromopyridine
SMILES C1=CC(=NC(=C1)Br)Br

Specifications

Melting Point 117°C to 120°C
Boiling Point 255°C
Flash Point 213°C (415°F)
Odor Recognizable
Quantity 100 g
UN Number UN2811
Beilstein 108922
Solubility Information Soluble in hot methanol. Insoluble in water.
Formula Weight 236.9
Percent Purity 98%
Chemical Name or Material 2,6-Dibromopyridine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10100683 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15397.22,A15397.14,A15397.36
Lot Number10160056 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15397.22,A15397.14,A15397.36
Lot Number10199314 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15397.22,A15397.14,A15397.36
Lot Number10182182 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15397.22,A15397.14,A15397.36
Lot NumberH01S030 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15397.22,A15397.14,A15397.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dibromopyridine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.