missing translation for 'onlineSavingsMsg'
Learn More
2,6-Dimethoxypyridine-3-boronic acid, 97%, Thermo Scientific™ Product Code.: 11986471

2,6-Dimethoxypyridine-3-boronic acid, 97%, Thermo Scientific™

Product Code. 11986471
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11986471

Brand: Thermo Scientific Acros 445410050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 221006-70-8
Molecular Formula C7H10BNO4
Molecular Weight (g/mol) 182.97
InChI Key ADGHSWFUZUADDH-UHFFFAOYSA-N
Synonym 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridine-5-boronic acid, boronic acid, 2,6-dimethoxy-3-pyridinyl, 2,6-dimethoxy-3-pyridyl boronic acidShow MoreShow Less
PubChem CID 2762707
IUPAC Name (2,6-dimethoxypyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1)OC)OC)(O)O

Specifications

Melting Point 108°C to 109°C
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 182.97
Percent Purity 97%
Chemical Name or Material 2, 6-Dimethoxypyridine-3-boronic acid
compliance-icons
Product Identifier
  • 2,6-Dimethoxypyridine-3-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dimethoxypyridine-3-boronic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.