missing translation for 'onlineSavingsMsg'
Learn More
2,6-Dimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals Product Code.: 11400478

2,6-Dimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11400478
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11400478

Brand: Thermo Scientific Alfa Aesar B24613.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for palladium catalyzed Suzuki-Miyaura coupling reactions, Pd-catalyzed homocouplings, Rhodium(I)-catalyzed 1,4-addition reactions. It is used in the preparation of protein kinase inhibitors, G Protein-Coupled Receptor.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100379-00-8
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD01009693
InChI Key ZXDTWWZIHJEZOG-UHFFFAOYSA-N
Synonym 2,6-dimethylphenyl boronic acid, 2,6-dimethylbenzeneboronic acid, 2,6-dimethylphenyl boranediol, 2,6-dimethylboronic acid, m-xylene-2-boronic acid, 2,6-dimethyl-phenylboronic acid, boronic acid, 2,6-dimethylphenyl, 2-borono-m-xylene, pubchem16357, acmc-1c3za
PubChem CID 583322
IUPAC Name (2,6-dimethylphenyl)boronic acid
SMILES CC1=CC=CC(C)=C1B(O)O

Specifications

Melting Point 120°C to 126°C (decomposition)
Quantity 25 g
Solubility Information Soluble in methanol.
Formula Weight 149.99
Percent Purity 97%
Chemical Name or Material 2,6-Dimethylbenzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5003A17R Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24613.06,B24613.03,B24613.14
Lot Number5005P24A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24613.06,B24613.03,B24613.14
Lot Number10145064 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24613.06,B24613.03,B24613.14
Lot Number10145228 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24613.06,B24613.03,B24613.14
Lot Number10097519 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24613.06,B24613.03,B24613.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dimethylbenzeneboronic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.