Learn More
2,6-Lutidine N-oxide, 98%, Thermo Scientific Chemicals
Description
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,6-Lutidine N-oxide is used as a ligand and form coordination complexes with metal salts.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with oxidizing agents, moisture and water.

Specifications
Specifications
Density | 1.102 |
Boiling Point | 108°C to 111°C (10 mmHg) |
Refractive Index | 1.5685 |
Quantity | 10 g |
Beilstein | 110551 |
Sensitivity | Hygroscopic |
Solubility Information | Miscible with water. |
Formula Weight | 123.16 |
Percent Purity | 98% |
Chemical Name or Material | 2,6-Lutidine N-oxide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10236225 | Certificate of Analysis | 30/08/2024 | A14299.30,A14299.09,A14299.18 |
10231521 | Certificate of Analysis | 12/01/2024 | A14299.30,A14299.09,A14299.18 |
10210939 | Certificate of Analysis | 12/01/2024 | A14299.30,A14299.09,A14299.18 |
10236225 | Certificate of Analysis | 22/11/2021 | A14299.30,A14299.09,A14299.18 |
10113177 | Certificate of Analysis | 22/11/2021 | A14299.30,A14299.09,A14299.18 |
Safety and Handling

- 2,6-Lutidine N-oxide
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 2,6-Lutidine N-oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.