Learn More
2-Acetamido-5-chlorothiazole, 97%, Thermo Scientific™
Product Code. 11373976
Click to view available options
Quantity:
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20256-39-7
Molecular Formula C5H5ClN2OS
Molecular Weight (g/mol) 176.618
MDL Number MFCD01318098
InChI Key YLRXYIGXXMJPOP-UHFFFAOYSA-N
Synonym 2-acetamido-5-chlorothiazole, n-5-chlorothiazol-2-yl acetamide, n-5-chloro-1,3-thiazol-2-yl acetamide, acmc-20ao6g, 2-acetamido-5-chlorthiazol, 2-acetylamino-5-chlorothiazole, n-5-chloro-2-thiazolyl acetamide, acetamide,n-5-chloro-2-thiazolyl, n-5-chloranyl-1,3-thiazol-2-yl ethanamide
PubChem CID 4684134
IUPAC Name N-(5-chloro-1,3-thiazol-2-yl)acetamide
SMILES CC(=O)NC1=NC=C(S1)Cl
Specifications
Melting Point 208°C to 212°C
Quantity 1 g
Beilstein 120475
Formula Weight 176.63
Percent Purity 97%
Chemical Name or Material 2-Acetamido-5-chlorothiazole