missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-5-methylbenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11339699
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13091-43-5
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00051705
InChI Key LCMZECCEEOQWLQ-UHFFFAOYSA-N
PubChem CID 2774400
IUPAC Name 2-amino-3-bromo-5-methylbenzoic acid
SMILES CC1=CC(Br)=C(N)C(=C1)C(O)=O

Specifications

Melting Point 202°C to 206°C
Quantity 1 g
Beilstein 3540992
Synonym 3-bromo-5-methylanthranilic acid, 2-amino-3-bromo-5-methyl-benzoic acid, 6-amino-5-bromo-m-toluic acid, benzoic acid, 2-amino-3-bromo-5-methyl, acmc-20ao5e, 4-amino-3-bromo-5-carboxytoluene, 2-bromo-6-carboxy-4-methylaniline, benzoic acid,2-amino-3-bromo-5-methyl, 2-azanyl-3-bromanyl-5-methyl-benzoic acid, 2-amino-3-bromo-5-methylbenzoic acid
Formula Weight 230.07
Percent Purity ≥98%
Chemical Name or Material 2-Amino-3-bromo-5-methylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'