missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitro-4-picoline, 98%, Thermo Scientific™

Product Code. 10123641
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6635-86-5
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00006315
InChI Key IKMZGACFMXZAAT-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-3-nitropyridine, 2-amino-3-nitro-4-picoline, 2-pyridinamine, 4-methyl-3-nitro, 2-amino-3-nitro-4-methylpyridine, 4-methyl-3-nitro-pyridin-2-ylamine, 4-methyl-3-nitro-2-pyridylamine, 4-methyl-3-nitro-2-pyridinamine, 4-methyl-3-nitro-2-aminopyridine, 4-methyl-3-nitropyridin-2-amin, 4mrz
PubChem CID 243166
IUPAC Name 4-methyl-3-nitropyridin-2-amine
SMILES CC1=C(C(=NC=C1)N)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 136°C to 141°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 153.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-nitro-4-picoline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'